5-Hydroxynicotinonitrile 96%

Supplier: Ambeed
Warning

A371374-1G A371374-25G A371374-10G A371374-250MG A371374-5G
103826-480EA 9 USD
103826-480 76840-748 76840-750 77101-582 77101-584 77101-586
5-Hydroxynicotinonitrile 96%
5-Hydroxynicotinonitrile

Precaución: for Research Use Only

Formula: C6H4N2O
MW: 120.11 g/mol
Boiling Pt: 394.7 °C (760 mm Hg)
Melting Pt: 213…217 °C
Storage Temperature: Ambient
MDL Number: MFCD11044307
CAS Number: 152803-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR