5-Hydroxynicotinaldehyde 95%

Supplier: Ambeed
Warning

A183043-250MG A183043-1G A183043-100MG
103815-594EA 171 USD
103815-594 103846-158 76813-640 77301-810 77611-780
5-Hydroxynicotinaldehyde 95%
5-Hydroxynicotinaldehyde

Precaución: for Research Use Only

Formula: C₆H₅NO₂
Boiling Pt: 391.9 °C (760 mm Hg)
Flash Pt: 190.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD13189119
CAS Number: 1060804-48-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR