5-Hydroxyisoindoline-1,3-dione 98%

Supplier: Ambeed
Warning

A333160-1G A333160-25G A333160-5G A333160-100MG A333160-10G A333160-250MG
103825-624EA 51 USD
103825-624 103836-588 76839-592 76839-594 76839-596 77032-904 77032-906 77032-908
5-Hydroxyisoindoline-1,3-dione 98%
5-Hydroxyisoindoline-1,3-dione

Precaución: for Research Use Only

Formula: C₈H₅NO₃
Storage Temperature: Ambient
MDL Number: MFCD11101027
CAS Number: 50727-06-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR