5-Hydroxyisobenzofuran-1,3-dione 95%

Supplier: Ambeed
Warning

A114888-100MG A114888-250MG A114888-1G A114888-5G A114888-25G
103798-498EA 15 USD
103798-498 103824-340 77284-420 77284-422 77311-156 77311-158 77747-841
5-Hydroxyisobenzofuran-1,3-dione 95%
5-Hydroxyisobenzofuran-1,3-dione

Precaución: for Research Use Only

Formula: C₈H₄O₄
Boiling Pt: 407.8±28.0 °C (760 mm Hg)
Melting Pt: 171…173 °C
Flash Pt: 179.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00997252
CAS Number: 27550-59-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR