5-Hydroxybenzofuran-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A173575-100MG A173575-250MG A173575-1G
103806-830EA 152 USD
103806-830 103845-130 76974-136 76974-138 77082-378
5-Hydroxybenzofuran-2-carboxylic acid 95%
5-Hydroxybenzofuran-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆O₄
Boiling Pt: 396.4±22.0 °C (760 mm Hg)
Melting Pt: 252…254 °C
Flash Pt: 193.5 °C
Storage Temperature: Ambient
MDL Number: MFCD11101014
CAS Number: 56172-36-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR