3,4-Dihydro-5-hydroxy-1H-quinolin-2-one 97%

Supplier: Ambeed
Warning

Synonyms: 5-Hydroxy-2-oxo-1,2,3,4-tetrahydroquinoline, 5-Hydroxy-3,4-dihydro-2(1H)-quinolinone

A250283-250MG A250283-1G A250283-5G
103811-918EA 17 USD
103811-918 103851-756 76771-642 77091-106 77091-108
3,4-Dihydro-5-hydroxy-1H-quinolin-2-one 97%
3,4-Dihydro-5-hydroxy-1H-quinolin-2-one

Precaución: for Research Use Only

Formula: C₉H₉NO₂
MW: 163.18 g/mol
Boiling Pt: 381.2 °C (760 mm Hg)
Melting Pt: 225…227 °C
Flash Pt: 184.3 °C
Storage Temperature: Ambient
MDL Number: MFCD01862194
CAS Number: 30389-33-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR