5-Hydroxy-2-nitrobenzoic acid 98%

Supplier: Ambeed
Warning

A131901-10G A131901-250MG A131901-1G A131901-25G A131901-5G
103801-654EA 6 USD
103801-654 76811-222 76811-224 77314-184 77314-186 77314-188
5-Hydroxy-2-nitrobenzoic acid 98%
5-Hydroxy-2-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅NO₅
Boiling Pt: 455 °C (760 mm Hg)
Melting Pt: 168…172 °C
Flash Pt: 209.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00017566
CAS Number: 610-37-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR