5-Hydroxy-2-nitrobenzaldehyde 98%

Supplier: Ambeed
Warning

A195853-1G A195853-10G A195853-25G A195853-5G
103816-790EA 9 USD
103816-790 103847-400 77085-348 77139-240 77139-242 77302-482
5-Hydroxy-2-nitrobenzaldehyde 98%
5-Hydroxy-2-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₅NO₄
Boiling Pt: 373 °C (760 mm Hg)
Melting Pt: 165…169 °C
Flash Pt: 173.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00007332
CAS Number: 42454-06-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR