6-Methylpyridin-3-ol 97%

Supplier: Ambeed
Warning

Synonyms: 2-Methylpyridin-5-ol, 3-Hydroxy-6-methylpyridine, 5-Hydroxy-2-methylpyridine

A246864-5G A246864-100G A246864-10G A246864-500G
103851-570EA 14 USD
103851-570 77090-870 77113-362 77113-364 77113-366
6-Methylpyridin-3-ol 97%
6-Methylpyridin-3-ol

Precaución: for Research Use Only

Formula: C₆H₇NO
MW: 109.13 g/mol
Melting Pt: 168…170 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006339
CAS Number: 1121-78-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR