5-Hydroxy-o-toluic acid 98%

Supplier: Ambeed
Warning

Synonyms: 5-Hydroxy-2-methylbenzoic acid

A179433-500G A179433-1G A179433-5G A179433-10G A179433-100G
103807-428EA 6 USD
103807-428 103845-774 76769-508 77083-226 77083-228 77598-238 77625-848
5-Hydroxy-o-toluic acid 98%
5-Hydroxy-o-toluic acid

Precaución: for Research Use Only

Formula: C₈H₈O₃
MW: 152.15 g/mol
Melting Pt: 182…187 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09833184
CAS Number: 578-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR