5-Hydroxy-2-methoxybenzaldehyde 98%

Supplier: Ambeed
Warning

A132920-10G A132920-250MG A132920-1G
103801-804EA 37 USD
103801-804 76996-488 76996-490 77314-390
5-Hydroxy-2-methoxybenzaldehyde 98%
5-Hydroxy-2-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₈O₃
Melting Pt: 110…113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03820046
CAS Number: 35431-26-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR