5-Hydroxy-1-indanone 97%

Supplier: Ambeed
Warning

A116096-100G A116096-10G A116096-25G A116096-1G A116096-5G
103798-672EA 8 USD
103798-672 103824-536 76687-856 76687-858 76687-860 77297-418 77311-358
5-Hydroxy-1-indanone 97%
5-Hydroxy-1-indanone

Precaución: for Research Use Only

Formula: C9H8O2
MW: 148.1600 g/mol
Boiling Pt: 343.8 °C (760 mm Hg)
Melting Pt: 175 °C
Flash Pt: 146.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00857527
CAS Number: 3470-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR