5-Hydroxy-1H-pyrazole-3-carboxylic acid 98%

Supplier: Ambeed
Warning

A130375-1G A130375-250MG
103801-416EA 68 USD
103801-416 77313-850 77683-158
5-Hydroxy-1H-pyrazole-3-carboxylic acid 98%
5-Hydroxy-1H-pyrazole-3-carboxylic acid

Precaución: For research use only.

Formula: C₄H₄N₂O₃
MW: 128.086 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12195917
CAS Number: 89603-60-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR