5H-Pyrrolo[2,3-b]pyrazine 98%
Supplier: Ambeed
|
Warning
|
A159177-100MG
A159177-250MG
A159177-1G
A159177-5G
103805-228EA
106
USD
103805-228
103843-444
77286-392
77286-394
77319-320
77319-322
5H-Pyrrolo[2,3-b]pyrazine 98%
5H-Pyrrolo[2,3-b]pyrazine
Precaución: for Research Use Only
Formula:
C₆H₅N₃ MW: 119.124 g/mol Boiling Pt: 236 °C (760 mm Hg) Melting Pt: 155…156 °C Flash Pt: 96 °C Storage Temperature: Ambient |
MDL Number:
MFCD09834819 CAS Number: 4745-93-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...