5-Hexynoic acid 97%
Supplier: Ambeed
|
Danger
|
A166209-10G
A166209-250MG
A166209-1G
A166209-25G
A166209-5G
103806-024EA
11
USD
103806-024
103844-284
76973-804
76973-806
77320-522
77320-524
77320-526
5-Hexynoic acid 97%
5-Hexynoic acid
Precaución: for Research Use Only
Formula:
C₆H₈O₂ Boiling Pt: 220.6 °C (760 mm Hg) Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00066346 CAS Number: 53293-00-8 UN: 3265 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...