5-Hexen-1-ol 97%

Supplier: Ambeed
Danger

A778791-1G A778791-5G A778791-10G A778791-100G
103840-968EA 16 USD
103840-968 76845-228 76845-230 77573-508 77599-068
5-Hexen-1-ol 97%
5-Hexen-1-ol

Precaución: for Research Use Only

Formula: C₆H₁₂O
Boiling Pt: 78 °C at 25 mmHg
Flash Pt: 47 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002981
CAS Number: 821-41-0
UN: 1987
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR