5H-Cyclopenta[b]pyridin-7(6H)-one 98%

Supplier: Ambeed
Warning

A215283-100MG A215283-250MG
103818-474EA 36 USD
103818-474 103849-178 76691-604 77600-738
5H-Cyclopenta[b]pyridin-7(6H)-one 98%
5H-Cyclopenta[b]pyridin-7(6H)-one

Precaución: for Research Use Only

Formula: C8H7NO
MW: 133.1500 g/mol
Boiling Pt: 267.4 °C (760 mm Hg)
Flash Pt: 122.62 °C
Storage Temperature: Freezer
MDL Number: MFCD07778341
CAS Number: 31170-78-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR