5H-Benzo[b]carbazole 95%

Supplier: Ambeed
Warning

A341771-1G A341771-5G A341771-100MG A341771-250MG
103825-844EA 342 USD
103825-844 103836-818 76839-848 76839-850 77251-410 77251-412
5H-Benzo[b]carbazole 95%
5H-Benzo[b]carbazole

Precaución: for Research Use Only

Formula: C₁₆H₁₁N
MW: 217.26 g/mol
Boiling Pt: 456.1 °C (760 mm Hg)
Flash Pt: 207.9 °C
Storage Temperature: Ambient
MDL Number: MFCD02316988
CAS Number: 243-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR