5-Formyl-2-thiophenecarboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 5-Formylthenoic Acid, 5-Formylthiophene-2-carboxylic acid

A263847-1G A263847-250MG A263847-10G
103812-642EA 28 USD
103812-642 103852-534 77092-178 77289-278 77748-501
5-Formyl-2-thiophenecarboxylic acid 97%
5-Formyl-2-thiophenecarboxylic acid

Precaución: for Research Use Only

Formula: C₆H₄O₃S
Boiling Pt: 371.4 °C (760 mm Hg)
Flash Pt: 178.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00191437
CAS Number: 4565-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR