5-Formylpicolinonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 5-Formylpyridine-2-carbonitrile

A173891-100MG A173891-10G A173891-250MG A173891-1G
103806-880EA 43 USD
103806-880 103845-176 77029-202 77029-204 77029-206 77082-464
5-Formylpicolinonitrile 97%
5-Formylpicolinonitrile

Precaución: for Research Use Only

Formula: C7H4N2O
MW: 132.1200 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12755972
CAS Number: 131747-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR