5-Formyl-2-furancarboxylic acid 98%

Supplier: Ambeed
Warning

A141726-10G A141726-5G A141726-1G A141726-100MG A141726-250MG
103803-050EA 11 USD
103803-050 77095-458 77299-242 77316-228 77611-292 77900-817
5-Formyl-2-furancarboxylic acid 98%
Formula: C₆H₄O₄
Boiling Pt: 356.9 °C (760 mm Hg)
Melting Pt: 209 °C
Flash Pt: 169.6 °C
Storage Temperature: Freezer
MDL Number: MFCD00020924
CAS Number: 13529-17-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR