5-Formyl-2-methoxybenzenesulfonamide 97%

Supplier: Ambeed
Warning

A101009-100MG A101009-250MG A101009-50MG A101009-1G
103791-916EA 63 USD
103791-916 103795-752 103821-444 77233-066 77233-068 77233-070 77282-640
5-Formyl-2-methoxybenzenesulfonamide 97%
5-Formyl-2-methoxybenzenesulfonamide

Precaución: for Research Use Only

Formula: C₈H₉NO₄S
MW: 215.23 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD25955005
CAS Number: 105764-07-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR