5-Fluorouracil 98%

Supplier: Ambeed
Danger

Synonyms: Fluorouracil, 5-Fluoropyrimidine-2,4(1H,3H)-dione, 5-FU

A370752-25G A370752-10G A370752-5G A370752-100G
103837-478EA 5 USD
103837-478 76840-726 77290-876 77306-814 77616-638
5-Fluorouracil 98%
5-Fluorouracil

Precaución: for Research Use Only

Formula: C₄H₃FN₂O₂
MW: 130.08 g/mol
Boiling Pt: 401.4 °C (760 mm Hg)
Melting Pt: 278…286 °C
Flash Pt: 196.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00006018
CAS Number: 51-21-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR