5-Fluorothiophene-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A178753-250MG A178753-1G A178753-5G A178753-100MG
103807-364EA 209 USD
103807-364 103845-704 76974-342 77083-140 77083-142 77671-530
5-Fluorothiophene-2-carboxylic acid 97%
5-Fluorothiophene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₃FO₂S
Boiling Pt: 271.2 °C (760 mm Hg)
Melting Pt: 139…141 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07787557
CAS Number: 4377-58-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR