2-Amino-5-fluoro-1,3-thiazole hydrochloride 97%

Supplier: Ambeed
Warning

A189677-100MG A189677-10G A189677-250MG A189677-1G A189677-5G
103816-216EA 66 USD
103816-216 103846-810 76769-852 76769-854 76769-856 77084-582 77084-584
2-Amino-5-fluoro-1,3-thiazole hydrochloride 97%
2-Amino-5-fluoro-1,3-thiazole hydrochloride

Precaución: for Research Use Only

Formula: C₃H₃FN₂S·ClH
Boiling Pt: 221.8 °C (760 mm Hg)
Flash Pt: 88 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09056854
CAS Number: 745053-64-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR