5-Fluoro-2-pyrimidinecarbonitrile 97%

Supplier: Ambeed
Warning

A162356-100MG A162356-250MG
103805-606EA 60 USD
103805-606 103843-856 77273-362 77273-364
5-Fluoro-2-pyrimidinecarbonitrile 97%
5-Fluoro-2-pyrimidinecarbonitrile

Precaución: for Research Use Only

Formula: C5H2FN3
Boiling Pt: 255.7 °C (760 mm Hg)
Flash Pt: 108.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11846243
CAS Number: 38275-55-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR