5-Fluoropyrimidin-2(1H)-one 98%

Supplier: Ambeed
Warning

A178000-100MG A178000-250MG A178000-1G
103807-288EA 31 USD
103807-288 103845-628 76784-484 76784-486 77083-026
5-Fluoropyrimidin-2(1H)-one 98%
5-Fluoropyrimidin-2(1H)-one

Precaución: for Research Use Only

Formula: C₄H₃FN₂O
MW: 114.07 g/mol
Melting Pt: 170…174 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00223678
CAS Number: 2022-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR