5-Fluoro-2,3-diaminopyridine 98%

Supplier: Ambeed
Warning

Synonyms: 5-Fluoropyridine-2,3-diamine

A130436-100MG A130436-250MG
103801-422EA 412 USD
103801-422 76972-036 76972-038
5-Fluoro-2,3-diaminopyridine 98%
5-Fluoro-2,3-diaminopyridine

Precaución: for Research Use Only

Formula: C₅H₆FN₃
MW: 127.12 g/mol
Boiling Pt: 303.9 °C (760 mm Hg)
Flash Pt: 137.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08062986
CAS Number: 212268-13-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR