3-Fluoro-5-hydroxypyridine 98%

Supplier: Ambeed
Warning

Synonyms: 3-Fluoro-5-pyridinol, 5-Fluoropyridin-3-ol

A124336-100G A124336-10G A124336-250MG A124336-25G A124336-1G A124336-5G
103800-518EA 15 USD
103800-518 76996-002 76996-004 76996-006 76996-008 77297-984 77312-656
3-Fluoro-5-hydroxypyridine 98%
3-Fluoro-5-hydroxypyridine

Precaución: for Research Use Only

Formula: C₅H₄FNO
Boiling Pt: 303 °C (760 mm Hg)
Flash Pt: 137 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03095199
CAS Number: 209328-55-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR