5-Fluoro-2-formylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 5-Fluoropyridine-2-carboxaldehyde, 5-Fluoropicolinaldehyde

A177099-1G A177099-250MG A177099-25G A177099-10G
103807-212EA 17 USD
103807-212 103845-538 77082-918 77138-584 77301-496 77550-134
5-Fluoro-2-formylpyridine 98%
5-Fluoro-2-formylpyridine

Precaución: for Research Use Only

Formula: C₆H₄FNO
MW: 125.1 g/mol
Boiling Pt: 173.6 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD07781242
CAS Number: 31181-88-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR