5-Fluoronicotinonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 3-Cyano-5-fluoropyridine

A812653-10G A812653-250MG
103841-098EA 114 USD
103841-098 76653-682 76653-684
5-Fluoronicotinonitrile 97%
5-Fluoronicotinonitrile

Precaución: for Research Use Only

Formula: C₆H₃FN₂
MW: 122.10 g/mol
Boiling Pt: 182.2 °C (760 mm Hg)
Melting Pt: 50…54 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09027295
CAS Number: 696-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR