5-Fluoronicotinic acid 98%

Supplier: Ambeed
Warning

A212213-1G A212213-5G
103818-174EA 8 USD
103818-174 103848-874 77087-324 77087-326
5-Fluoronicotinic acid 98%
5-Fluoronicotinic acid

Precaución: for Research Use Only

Formula: C₆H₄FNO₂
MW: 141.10 g/mol
Boiling Pt: 272.2 °C (760 mm Hg)
Melting Pt: 193…198 °C
Flash Pt: 118.4 °C
Storage Temperature: Ambient
MDL Number: MFCD01318537
CAS Number: 402-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR