3-Fluoro-5-formylpyridine 97%

Supplier: Ambeed
Warning

Synonyms: 5-Fluoronicotinaldehyde

A120690-100G A120690-25G A120690-1G A120690-5G A120690-10G
103799-980EA 13 USD
103799-980 77094-440 77094-442 77297-778 77312-090 77571-230
3-Fluoro-5-formylpyridine 97%
3-Fluoro-5-formylpyridine

Precaución: for Research Use Only

Formula: C₆H₄FNO
MW: 125.1 g/mol
Boiling Pt: 188.6 °C (760 mm Hg)
Melting Pt: 38…41 °C
Flash Pt: 68 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07778429
CAS Number: 39891-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR