5-Fluoroisoindoline hydrochloride 97%

Supplier: Ambeed
Warning

A195968-100MG A195968-250MG A195968-1G A195968-5G A195968-10G
103816-800EA 46 USD
103816-800 103847-414 76814-094 76814-096 77085-370 77302-490 77671-740
5-Fluoroisoindoline hydrochloride 97%
5-Fluoroisoindoline hydrochloride

Precaución: for Research Use Only

Formula: C8H9ClFN
Boiling Pt: 235.3 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD09832541
CAS Number: 685565-15-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR