5-Fluorocytosine 97%

Supplier: Ambeed
Warning

Synonyms: 5-FC, 4-Amino-5-fluoro-2-pyrimidinol, Flucytosine, 6-Amino-5-fluoropyrimidin-2(1H)-one

A200898-1000G A200898-100G A200898-10G A200898-500G A200898-25G A200898-5G
103847-886EA 5 USD
103847-886 77274-832 77274-834 77274-836 77274-838 77302-730 77302-732
5-Fluorocytosine 97%
5-Fluorocytosine

Precaución: for Research Use Only

Formula: C₄H₄FN₃O
MW: 129.09 g/mol
Boiling Pt: 298
Melting Pt: 165
Storage Temperature: Ambient
MDL Number: MFCD00006035
CAS Number: 2022-85-7
Merck Index: 13,04151

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR