5-Fluoro-6-methylpyridine-2-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
A234611-100MG
A234611-250MG
A234611-25G
A234611-1G
103810-964EA
113
USD
103810-964
103850-720
77031-146
77031-148
77031-150
77303-988
5-Fluoro-6-methylpyridine-2-carboxylic acid 97%
5-Fluoro-6-methylpyridine-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₇H₆FNO₂ MW: 155.126 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11040267 CAS Number: 1005474-88-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...