5-Fluoro-6-methoxynicotinic acid 95%

Supplier: Ambeed
Warning

A198649-100MG A198649-10G A198649-250MG A198649-1G A198649-5G
103817-040EA 28 USD
103817-040 103847-668 77030-070 77030-072 77030-074 77085-690 77085-692
5-Fluoro-6-methoxynicotinic acid 95%
5-Fluoro-6-methoxynicotinic acid

Precaución: for Research Use Only

Formula: C₇H₆FNO₃
Storage Temperature: Ambient
MDL Number: MFCD11040262
CAS Number: 953780-42-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR