2-Amino-5-fluoro-3-methylpyridine 98%

Supplier: Ambeed
Warning

A155393-100MG A155393-250MG
103804-752EA 35 USD
103804-752 103842-948 77010-972 77010-974
2-Amino-5-fluoro-3-methylpyridine 98%
2-Amino-5-fluoro-3-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₇FN₂
Boiling Pt: 215.4 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD07375130
CAS Number: 886365-56-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR