5-Fluoro-3-methoxypyridin-2-amine 95%

Supplier: Ambeed
Warning

A444992-100MG A444992-50MG A444992-5G A444992-1G A444992-250MG
103827-722EA 293 USD
103827-722 77168-834 77168-836 77168-838 77307-338 77613-368
5-Fluoro-3-methoxypyridin-2-amine 95%
5-Fluoro-3-methoxypyridin-2-amine

Precaución: for Research Use Only

Formula: C₆H₇FN₂O
MW: 142.131 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18256739
CAS Number: 1097264-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR