5-Fluoropicolinic acid 98%

Supplier: Ambeed
Warning

Synonyms: 5-Fluoro-2-pyridinecarboxylic acid, 5-Fluoro-2-picolinic acid, 5-Fluoropyridine-2-carboxylic acid

A121376-100G A121376-10G A121376-250MG A121376-25G A121376-1G A121376-5G
103800-066EA 15 USD
103800-066 76766-502 76766-504 76766-506 76766-508 77312-182 77312-184
5-Fluoropicolinic acid 98%
5-Fluoropicolinic acid

Precaución: for Research Use Only

Formula: C₆H₄FNO₂
MW: 141.1 g/mol
Boiling Pt: 275.1 °C (760 mm Hg)
Melting Pt: 164…166 °C
Flash Pt: 120.2 °C
Storage Temperature: Ambient
MDL Number: MFCD04114196
CAS Number: 107504-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR