5-Fluoro-2-nitropyridine 97%

Supplier: Ambeed
Warning

A225131-1G A225131-25G A225131-5G A225131-10G A225131-250MG
103819-160EA 14 USD
103819-160 103850-052 77088-722 77088-724 77088-726 77139-914 77139-916
5-Fluoro-2-nitropyridine 97%
5-Fluoro-2-nitropyridine

Precaución: for Research Use Only

Formula: C5H3FN2O2
Storage Temperature: Ambient
MDL Number: MFCD03092917
CAS Number: 779345-37-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR