5-Fluoro-2-nitrobenzonitrile 97%
Supplier: Ambeed
|
Warning
|
A409820-1G
A409820-25G
A409820-10G
103827-202EA
6
USD
103827-202
103838-250
76773-796
76841-762
76841-764
77626-824
5-Fluoro-2-nitrobenzonitrile 97%
5-Fluoro-2-nitrobenzonitrile
Precaución: for Research Use Only
Formula:
C₇H₃FN₂O₂ Boiling Pt: 302.9 °C (760 mm Hg) Melting Pt: 84…88 °C Flash Pt: 137 °C Storage Temperature: Ambient |
MDL Number:
MFCD01632196 CAS Number: 50594-78-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...