5-Fluoro-2-nitrobenzenesulfonyl chloride 97%, Ambeed.Inc

Supplier: Ambeed
Danger

Synonyms: 5-Fluoro-2-nitrobenzene-1-sulphonyl chloride

A565880-5G A565880-100MG A565880-250MG
103828-772EA 90 USD
103828-772 103839-912 76844-026 77169-846 77684-420
5-Fluoro-2-nitrobenzenesulfonyl chloride 97%, Ambeed.Inc
5-Fluoro-2-nitrobenzenesulfonyl chloride

Precaución: For research use only.

Formula: C₆H₃ClFNO₄S
MW: 239.609 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11858121
CAS Number: 82711-97-5
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR