5-Fluoro-2-nitrobenzamide 97%

Supplier: Ambeed
Warning

A300132-1G A300132-250MG
103814-262EA 49 USD
103814-262 103835-412 76838-100 77166-950
5-Fluoro-2-nitrobenzamide 97%
5-Fluoro-2-nitrobenzamide

Precaución: for Research Use Only

Formula: C₇H₅FN₂O₃
MW: 184.13 g/mol
Boiling Pt: 275.6±25.0 °C (760 mm Hg)
Melting Pt: 93…97 °C
Flash Pt: 120.5 °C
Storage Temperature: Ambient
MDL Number: MFCD11870104
CAS Number: 77206-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR