5-Fluoro-2-nitrobenzaldehyde 98%

Supplier: Ambeed
Warning

A272812-100G A272812-10G A272812-1G A272812-500G A272812-25G A272812-5G
103853-026EA 9 USD
103853-026 76772-166 76772-168 76772-170 76772-172 77092-812 77305-114
5-Fluoro-2-nitrobenzaldehyde 98%
5-Fluoro-2-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄FNO₃
MW: 169.11 g/mol
Boiling Pt: 287.6 °C (760 mm Hg)
Melting Pt: 92…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00153175
CAS Number: 395-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR