5-Fluoro-2-methylindole 95%

Supplier: Ambeed
Warning

A212318-10G A212318-25G A212318-1G
103818-190EA 9 USD
103818-190 103848-888 76691-542 76691-544 77303-242
5-Fluoro-2-methylindole 95%
5-Fluoro-2-methylindole

Precaución: for Research Use Only

Formula: C9H8FN
Boiling Pt: 269.2 °C (760 mm Hg)
Melting Pt: 98…101 °C
Flash Pt: 116.6 °C
Storage Temperature: Ambient
MDL Number: MFCD02093649
CAS Number: 399-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR