5-Fluoro-o-toluonitrile 98%

Supplier: Ambeed
Warning

A172063-10G A172063-1G A172063-25G A172063-100G
103844-972EA 10 USD
103844-972 76769-258 76769-260 77082-150 77625-784
5-Fluoro-o-toluonitrile 98%
5-Fluoro-o-toluonitrile

Precaución: for Research Use Only

Formula: C₈H₆FN
Melting Pt: 43…45 °C
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00042295
CAS Number: 77532-79-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR