5-Fluoro-2-methoxypyridin-3-amine 97%

Supplier: Ambeed
Warning

A140407-100MG A140407-10G A140407-250MG A140407-1G A140407-5G
103802-864EA 94 USD
103802-864 76823-404 76823-406 76823-408 77299-140 77315-980
5-Fluoro-2-methoxypyridin-3-amine 97%
5-Fluoro-2-methoxypyridin-3-amine

Precaución: for Research Use Only

Formula: C₆H₇FN₂O
MW: 142.131 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18261779
CAS Number: 1211541-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR