5-Fluoro-o-anisidine 98%

Supplier: Ambeed
Danger

Synonyms: 5-Fluoro-2-methoxyaniline, 5-Fluoro-2-anisidine, 2-Amino-4-fluoroanisole

A119506-100G A119506-10G A119506-25G A119506-1G A119506-5G
103799-778EA 5 USD
103799-778 76766-386 76766-388 76766-390 77311-894 77311-896
5-Fluoro-o-anisidine 98%
5-Fluoro-o-anisidine

Precaución: for Research Use Only

Formula: C₇H₈FNO
MW: 141.15 g/mol
Boiling Pt: 234.3 °C (760 mm Hg)
Melting Pt: 3 °C
Flash Pt: 108.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00077537
CAS Number: 1978-39-8
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR