5-Fluoro-2-iodopyridine 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Iodo-5-fluoropyridine
A238250-100MG
A238250-250MG
A238250-5G
103811-216EA
44
USD
103811-216
103850-998
76771-340
76771-342
77090-110
5-Fluoro-2-iodopyridine 95%
5-Fluoro-2-iodopyridine
Precaución: for Research Use Only
Formula:
C5H3FIN MW: 222.99 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD07787574 CAS Number: 159870-80-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...